Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tex2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_198292.3 | NP_938034.2 | 5 - 6 | 2532 | 75 | |
GenBank mRNA | AK129434.1 | - | 5 - 6 | 2532 | 75 | |
AK168397.1 | - | 5 - 6 | 2542 | 75 | ||
BC057406.1 | - | 4 - 5 | 942 | 5687258 | 75 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |