Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Otud7b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025613.1 | NP_001020784.1 | 8 - 9 | 1263 | 103 | |
NM_001025614.1 | NP_001020785.1 | 7 - 8 | 1205 | 103 | ||
GenBank mRNA | AK044701.1 | - | 8 - 9 | 1264 | 103 | |
BC009124.1 | - | 2 - 3 | 172 | 103 | ||
BC037040.1 | - | 10 - 11 | 1249 | 103 | ||
BC141397.1 | - | 7 - 8 | 1095 | 9056010 | 103 | |
BC141398.1 | - | 7 - 8 | 1095 | 9056011 | 103 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |