Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pde4dip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039376.2 | NP_001034465.2 | 39 - 40 | 6310 | 52 | |
NM_001289701.1 | NP_001276630.1 | 39 - 40 | 6361 | 52 | ||
NM_001289702.1 | NP_001276631.1 | 38 - 39 | 6208 | 52 | ||
GenBank mRNA | BC141185.1 | - | 39 - 40 | 6361 | 9055797 | 52 |
BC145504.1 | - | 38 - 39 | 6208 | 9054067 | 52 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |