Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccnyl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001097644.1 | NP_001091113.1 | 1 - 2 | 54 | 69 | |
GenBank mRNA | AK132782.1 | - | 409 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Mus musculus Probe spans exons |