Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prkab2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_182997.2 | NP_892042.2 | 3 - 4 | 501 | 80 | |
GenBank mRNA | BC060228.1 | - | 3 - 4 | 501 | 6839848 | 80 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protein Kinase Mammalian Gene Collection Mus musculus Probe spans exons |