Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gls | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081081.2 | NP_001074550.1 | 7 - 8 | 1297 | 73 | |
NM_001113383.1 | NP_001106854.1 | 7 - 8 | 1297 | 73 | ||
GenBank mRNA | AK046297.1 | - | 2 - 3 | 219 | 73 | |
AK048072.1 | - | 5 - 6 | 475 | 73 | ||
AK173039.1 | - | 7 - 8 | 1297 | 73 | ||
BC025534.1 | - | 1 - 2 | 81 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |