Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pstpip2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013831.4 | NP_038859.3 | 203 | 58 | ||
GenBank mRNA | AK033996.1 | - | 1 - 2 | 191 | 58 | |
AK132418.1 | - | 185 | 58 | |||
AK154284.1 | - | 170 | 58 | |||
AK154724.1 | - | 1 - 2 | 209 | 58 | ||
AK157994.1 | - | 1 - 2 | 187 | 58 | ||
BC002123.1 | - | 184 | 3494357 | 58 | ||
BC057654.1 | - | 145 | 4460817 | 58 | ||
CT010308.1 | - | 1 - 2 | 35 | 58 | ||
Y18101.1 | - | 1 - 2 | 35 | 58 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Probe spans exons |