Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plekhg3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_153804.4 | NP_722499.4 | 4 - 5 | 816 | 99 | |
GenBank mRNA | AK080928.1 | - | 4 - 5 | 817 | 99 | |
AK162025.1 | - | 4 - 5 | 758 | 99 | ||
BC030417.1 | - | 3 - 4 | 159 | 99 | ||
BC049264.1 | - | 3 - 4 | 148 | 99 | ||
BC096443.1 | - | 4 - 5 | 794 | 4189193 | 99 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |