Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pik3cd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001029837.2 | NP_001025008.2 | 21 - 22 | 3000 | 66 | |
NM_001164049.1 | NP_001157521.1 | 21 - 22 | 3027 | 66 | ||
NM_001164050.1 | NP_001157522.1 | 21 - 22 | 3021 | 66 | ||
NM_001164051.1 | NP_001157523.1 | 21 - 22 | 3048 | 66 | ||
NM_001164052.1 | NP_001157524.1 | 19 - 20 | 2742 | 66 | ||
NM_008840.3 | NP_032866.2 | 22 - 23 | 3193 | 66 | ||
GenBank mRNA | AK040867.1 | - | 21 - 22 | 2962 | 66 | |
AK149935.1 | - | 22 - 23 | 3092 | 66 | ||
AK171025.1 | - | 21 - 22 | 3007 | 66 | ||
AK172222.1 | - | 21 - 22 | 2904 | 66 | ||
BC035203.1 | - | 21 - 22 | 3011 | 4192906 | 66 | |
CT010349.1 | - | 19 - 20 | 2716 | 66 | ||
U86587.1 | - | 19 - 20 | 2710 | 66 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |