Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Apcdd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133237.3 | NP_573500.2 | 2 - 3 | 583 | 139 | |
GenBank mRNA | AK156319.1 | - | 2 - 3 | 584 | 139 | |
AK172004.1 | - | 587 | 139 | |||
BC141269.1 | - | 3 - 4 | 357 | 9055881 | 139 | |
BC141270.1 | - | 3 - 4 | 403 | 9055882 | 139 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |