Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prrc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028447.3 | NP_082723.2 | 3 - 4 | 664 | 73 | |
GenBank mRNA | AK004443.1 | - | 3 - 4 | 658 | 73 | |
AK143537.1 | - | 3 - 4 | 656 | 73 | ||
AK182391.1 | - | 1 - 2 | 121 | 73 | ||
BC014817.1 | - | 3 - 4 | 580 | 3709357 | 73 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Mus musculus Probe spans exons |