Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atp8b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001001488.3 | NP_001001488.2 | 13 - 14 | 1746 | 91 | |
GenBank mRNA | AK157316.1 | - | 13 - 14 | 2014 | 91 | |
AY506548.1 | - | 13 - 14 | 1475 | 91 | ||
BC117946.2 | - | 13 - 14 | 1473 | 40098035 | 91 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Transporter Mammalian Gene Collection Mus musculus Probe spans exons |