Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pias2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001164167.1 | NP_001157639.1 | 8 - 9 | 1227 | 85 | |
NM_001164168.1 | NP_001157640.1 | 8 - 9 | 1243 | 85 | ||
NM_001164169.1 | NP_001157641.1 | 8 - 9 | 1227 | 85 | ||
NM_001164170.1 | NP_001157642.1 | 8 - 9 | 1243 | 85 | ||
NM_008602.4 | NP_032628.3 | 8 - 9 | 1227 | 85 | ||
GenBank mRNA | AF039567.1 | - | 7 - 8 | 672 | 85 | |
AK014871.1 | - | 8 - 9 | 1218 | 85 | ||
AK029716.1 | - | 8 - 9 | 1228 | 85 | ||
AK078097.1 | - | 3 - 4 | 295 | 85 | ||
AK078813.1 | - | 8 - 9 | 1197 | 85 | ||
AK086653.1 | - | 8 - 9 | 1159 | 85 | ||
AK149142.1 | - | 9 - 10 | 1265 | 85 | ||
BC005596.1 | - | 8 - 9 | 1104 | 3969546 | 85 | |
BC034711.1 | - | 8 - 9 | 1233 | 2631738 | 85 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |