Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nfib | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113210.2 | NP_001106681.1 | 9 - 10 | 2408 | 88 | |
NM_001286127.1 | NP_001273056.1 | 9 - 10 | 2408 | 88 | ||
GenBank mRNA | AA500678.1 | - | 2 - 3 | 337 | 88 | |
BC023366.1 | - | 9 - 10 | 1545 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |