Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prdm6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033281.3 | NP_001028453.1 | 3 - 4 | 1045 | 152 | |
GenBank mRNA | AK133910.1 | - | 3 - 4 | 1046 | 152 | |
BC139295.1 | - | 3 - 4 | 1075 | 8862317 | 152 | |
DQ340564.1 | - | 3 - 4 | 621 | 152 | ||
DQ340565.1 | - | 3 - 4 | 540 | 152 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |