Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mtap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_024433.2 | NP_077753.1 | 1 - 2 | 162 | 99 | |
GenBank mRNA | AK005064.1 | - | 1 - 2 | 109 | 99 | |
AK011421.1 | - | 1 - 2 | 107 | 99 | ||
AK089916.1 | - | 1 - 2 | 96 | 99 | ||
AK167319.1 | - | 1 - 2 | 163 | 99 | ||
BC003858.1 | - | 1 - 2 | 106 | 3485422 | 99 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |