Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrp8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080926.1 | NP_001074395.1 | 3 - 4 | 502 | 60 | |
NR_033496.1 | 3 - 4 | 502 | 60 | |||
GenBank mRNA | AJ312058.1 | - | 3 - 4 | 348 | 60 | |
AK030143.1 | - | 1 - 2 | 137 | 60 | ||
AK038580.1 | - | 2 - 3 | 282 | 60 | ||
BC127061.1 | - | 4 - 5 | 342 | 60 | ||
D85463.1 | - | 3 - 4 | 500 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |