Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Btf3l4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027453.2 | NP_081729.1 | 3 - 4 | 252 | 128 | |
GenBank mRNA | AK004239.1 | - | 3 - 4 | 283 | 128 | |
AK011367.1 | - | 3 - 4 | 598 | 128 | ||
AK013120.1 | - | 3 - 4 | 240 | 128 | ||
AK076283.1 | - | 3 - 4 | 252 | 128 | ||
AK167256.1 | - | 3 - 4 | 233 | 128 | ||
AK210996.1 | - | 1 - 2 | 124 | 128 | ||
AK211852.1 | - | 1 - 2 | 124 | 128 | ||
BC024612.1 | - | 3 - 4 | 235 | 128 | ||
BC024920.1 | - | 3 - 4 | 195 | 128 | ||
BC058282.1 | - | 3 - 4 | 208 | 30077136 | 128 | |
BC099407.1 | - | 2 - 3 | 187 | 30938785 | 128 | |
BC138215.1 | - | 3 - 4 | 233 | 8861234 | 128 | |
BC138216.1 | - | 3 - 4 | 233 | 8861235 | 128 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |