Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Scp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011327.4 | NP_035457.1 | 5 - 6 | 498 | 133 | |
GenBank mRNA | AK004860.2 | - | 5 - 6 | 428 | 133 | |
BC018384.1 | - | 5 - 6 | 453 | 3710340 | 133 | |
BC034613.1 | - | 5 - 6 | 419 | 5123611 | 133 | |
M91458.1 | - | 5 - 6 | 413 | 133 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |