Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpr89 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026229.4 | NP_080505.1 | 1 - 2 | 117 | 70 | |
GenBank mRNA | AK002937.1 | - | 1 - 2 | 137 | 70 | |
AK016787.1 | - | 1 - 2 | 117 | 70 | ||
BC010729.1 | - | 1 - 2 | 110 | 3708143 | 70 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protein Kinase Mammalian Gene Collection Mus musculus Probe spans exons |