Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpr89 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026229.4 | NP_080505.1 | 9 - 10 | 889 | 150 | |
GenBank mRNA | AK009734.1 | - | 8 - 9 | 768 | 150 | |
AK016787.1 | - | 9 - 10 | 889 | 150 | ||
BC010729.1 | - | 9 - 10 | 882 | 3708143 | 150 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Protein Kinase Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |