Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gnal | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010307.3 | NP_034437.1 | 7 - 8 | 885 | 64 | |
NM_177137.5 | NP_796111.2 | 7 - 8 | 1050 | 64 | ||
GenBank mRNA | AK158831.1 | - | 7 - 8 | 886 | 64 | |
AY179169.1 | - | 7 - 8 | 619 | 64 | ||
BC078439.1 | - | 7 - 8 | 1012 | 5685967 | 64 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |