Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Diap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK156391.1 | - | 10 - 11 | 1437 | 66 | |
AK220466.1 | - | 23 - 24 | 3186 | 66 | ||
BC070412.1 | - | 24 - 25 | 3351 | 5705112 | 66 | |
U96963.1 | - | 22 - 23 | 3189 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |