Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc26a4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011867.3 | NP_035997.1 | 18 - 19 | 2295 | 109 | |
GenBank mRNA | AF167411.1 | - | 18 - 19 | 2296 | 109 | |
BC140995.1 | - | 17 - 18 | 2143 | 9055607 | 109 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Transporter Mammalian Gene Collection Mus musculus Probe spans exons |