Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hdac3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010411.2 | NP_034541.2 | 12 - 13 | 1051 | 84 | |
GenBank mRNA | AF074881.1 | - | 12 - 13 | 998 | 84 | |
AF074882.1 | - | 6 - 7 | 425 | 84 | ||
AF098295.1 | - | 12 - 13 | 976 | 84 | ||
AF125536.1 | - | 12 - 13 | 983 | 84 | ||
AK145158.1 | - | 12 - 13 | 1017 | 84 | ||
AK206656.1 | - | 5 - 6 | 338 | 84 | ||
AK208847.1 | - | 1 - 2 | 64 | 84 | ||
BC139300.1 | - | 12 - 13 | 1079 | 8862322 | 84 | |
BC139301.1 | - | 12 - 13 | 1079 | 8862323 | 84 | |
JN651903.1 | - | 6 - 7 | 408 | 84 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |