Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Afap1l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_178928.4 | NP_849259.2 | 8 - 9 | 1000 | 103 | |
GenBank mRNA | AK035227.1 | - | 8 - 9 | 1000 | 103 | |
AK053714.1 | - | 8 - 9 | 999 | 103 | ||
BC094505.1 | - | 5 - 6 | 438 | 103 | ||
BC138923.1 | - | 8 - 9 | 961 | 8861945 | 103 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |