Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppargc1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133249.2 | NP_573512.1 | 2 - 3 | 313 | 65 | |
GenBank mRNA | AF453324.1 | - | 2 - 3 | 313 | 65 | |
AK028464.1 | - | 3 - 4 | 363 | 65 | ||
AK086111.1 | - | 2 - 3 | 308 | 65 | ||
BC150699.1 | - | 2 - 3 | 293 | 9087610 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |