Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Osbpl9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001134791.2 | NP_001128263.1 | 17 - 18 | 1695 | 113 | |
NM_133885.4 | NP_598646.4 | 20 - 21 | 1928 | 113 | ||
NM_173350.3 | NP_775485.2 | 18 - 19 | 1734 | 113 | ||
GenBank mRNA | AK035658.1 | - | 18 - 19 | 1684 | 113 | |
AK075981.1 | - | 7 - 8 | 714 | 113 | ||
AK131984.1 | - | 20 - 21 | 1900 | 113 | ||
AK172010.1 | - | 18 - 19 | 1721 | 113 | ||
AK178534.1 | - | 3 - 4 | 244 | 113 | ||
AK202959.1 | - | 3 - 4 | 244 | 113 | ||
AK207117.1 | - | 3 - 4 | 245 | 113 | ||
BC002157.1 | - | 1 - 2 | 52 | 113 | ||
BC021507.1 | - | 16 - 17 | 1556 | 5347121 | 113 | |
BC023759.1 | - | 20 - 21 | 1883 | 113 | ||
BC026927.1 | - | 18 - 19 | 1617 | 5134780 | 113 | |
BC089163.1 | - | 17 - 18 | 1636 | 5367603 | 113 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |