Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Megf6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001162977.1 | NP_001156449.1 | 11 - 12 | 1672 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Mus musculus Probe spans exons |