Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lman1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001172062.1 | NP_001165533.1 | 10 - 11 | 1266 | 60 | |
NM_027400.3 | NP_081676.1 | 10 - 11 | 1266 | 60 | ||
GenBank mRNA | AK011495.1 | - | 10 - 11 | 1252 | 60 | |
AK151599.1 | - | 10 - 11 | 1253 | 60 | ||
AK151949.1 | - | 10 - 11 | 1253 | 60 | ||
AK153554.1 | - | 10 - 11 | 1253 | 60 | ||
BC057165.1 | - | 10 - 11 | 1275 | 4948301 | 60 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |