Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Malt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172833.2 | NP_766421.1 | 5 - 6 | 1025 | 116 | |
GenBank mRNA | AK041919.1 | - | 5 - 6 | 1030 | 116 | |
AK045200.1 | - | 5 - 6 | 1016 | 116 | ||
AK085071.1 | - | 5 - 6 | 1025 | 116 | ||
AK164626.1 | - | 5 - 6 | 1061 | 116 | ||
BC110487.1 | - | 5 - 6 | 999 | 40048907 | 116 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Apoptosis Protease Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |