Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdk11b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007661.3 | NP_031687.2 | 6 - 7 | 719 | 71 | |
GenBank mRNA | AK008283.1 | - | 6 - 7 | 696 | 71 | |
AK077668.1 | - | 6 - 7 | 720 | 71 | ||
AK135402.1 | - | 6 - 7 | 726 | 71 | ||
AK147133.1 | - | 6 - 7 | 697 | 71 | ||
BC052920.1 | - | 6 - 7 | 797 | 6489832 | 71 | |
L37092.1 | - | 5 - 6 | 1294 | 71 | ||
M58633.1 | - | 1 - 2 | 59 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |