Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnajb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001159883.1 | NP_001153355.1 | 8 - 9 | 818 | 66 | |
NM_001159884.1 | NP_001153356.1 | 8 - 9 | 766 | 66 | ||
NM_001159885.1 | NP_001153357.1 | 7 - 8 | 801 | 66 | ||
NM_020266.2 | NP_064662.2 | 7 - 8 | 764 | 66 | ||
NM_178055.4 | NP_835156.1 | 8 - 9 | 818 | 66 | ||
GenBank mRNA | AB028858.1 | - | 7 - 8 | 631 | 66 | |
AK076061.1 | - | 8 - 9 | 754 | 66 | ||
AK156083.1 | - | 8 - 9 | 787 | 66 | ||
AK171494.1 | - | 8 - 9 | 764 | 66 | ||
BC011090.1 | - | 8 - 9 | 720 | 3968468 | 66 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons |