Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prkag3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_153744.3 | NP_714966.1 | 1 - 2 | 145 | 103 | |
GenBank mRNA | AF525500.1 | - | 1 - 2 | 52 | 103 | |
AK036585.1 | - | 1 - 2 | 146 | 103 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Protein Kinase Detect Genomic DNA Mus musculus Probe spans exons |