Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sh3rf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001146299.1 | NP_001139771.1 | 5 - 6 | 1263 | 66 | |
NM_172966.3 | NP_766554.2 | 4 - 5 | 1167 | 66 | ||
GenBank mRNA | AK028153.1 | - | 4 - 5 | 663 | 66 | |
AK029168.1 | - | 4 - 5 | 1168 | 66 | ||
AK033601.1 | - | 5 - 6 | 1307 | 66 | ||
BC137955.1 | - | 5 - 6 | 1216 | 8860974 | 66 | |
BC145060.1 | - | 5 - 6 | 1216 | 9053608 | 66 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Mus musculus Inventoried Probe spans exons |