Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fkbp15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045528.1 | NP_001038993.1 | 19 - 20 | 2035 | 92 | |
GenBank mRNA | AK049459.1 | - | 19 - 20 | 1980 | 92 | |
AK049509.1 | - | 19 - 20 | 2035 | 92 | ||
AK122343.1 | - | 12 - 13 | 1274 | 92 | ||
AK161705.1 | - | 19 - 20 | 1965 | 92 | ||
BC034098.1 | - | 9 - 10 | 972 | 92 | ||
BC060651.1 | - | 19 - 20 | 1949 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |