Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrrc42 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_029985.2 | NP_084261.1 | 6 - 7 | 1194 | 104 | |
GenBank mRNA | AK020847.1 | - | 6 - 7 | 1000 | 104 | |
AK169814.1 | - | 4 - 5 | 652 | 104 | ||
BC027203.1 | - | 6 - 7 | 1126 | 3710091 | 104 | |
BC031894.1 | - | 6 - 7 | 1098 | 5362699 | 104 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |