Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kcnab2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001252654.1 | NP_001239583.1 | 6 - 7 | 487 | 80 | |
NM_001252655.1 | NP_001239584.1 | 6 - 7 | 516 | 80 | ||
NM_001252656.1 | NP_001239585.1 | 7 - 8 | 558 | 80 | ||
NM_010598.3 | NP_034728.2 | 7 - 8 | 529 | 80 | ||
GenBank mRNA | AK143166.1 | - | 6 - 7 | 464 | 80 | |
AK152613.1 | - | 7 - 8 | 481 | 80 | ||
AK157053.1 | - | 6 - 7 | 480 | 80 | ||
BC039178.1 | - | 7 - 8 | 459 | 4503373 | 80 | |
L48983.1 | - | 7 - 8 | 521 | 80 | ||
U65592.1 | - | 6 - 7 | 80 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Ion Channel Mammalian Gene Collection Mus musculus Probe spans exons |