Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Obox6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145710.2 | NP_663756.2 | 1 - 2 | 48 | 77 | |
GenBank mRNA | BY731792.1 | - | 1 - 2 | 47 | 77 | |
CN712286.1 | - | 1 - 2 | 48 | 77 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Detect Genomic DNA Mus musculus Probe spans exons |