Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmeff2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019790.4 | NP_062764.1 | 1 - 2 | 593 | 85 | |
GenBank mRNA | AB017270.1 | - | 1 - 2 | 556 | 85 | |
AB041565.1 | - | 1 - 2 | 597 | 85 | ||
AK029307.1 | - | 1 - 2 | 593 | 85 | ||
AK030053.1 | - | 6 - 7 | 1829 | 85 | ||
AK076254.1 | - | 1 - 2 | 593 | 85 | ||
AK135038.1 | - | 1 - 2 | 580 | 85 | ||
BC034850.1 | - | 1 - 2 | 569 | 1397175 | 85 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |