Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acsl5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027976.2 | NP_082252.1 | 14 - 15 | 2137 | 62 | |
GenBank mRNA | AK006541.1 | - | 3 - 4 | 614 | 62 | |
AK034942.1 | - | 2137 | 62 | |||
AK145894.1 | - | 14 - 15 | 1798 | 62 | ||
AK150542.1 | - | 14 - 15 | 1803 | 62 | ||
AK150663.1 | - | 14 - 15 | 1803 | 62 | ||
AK151378.1 | - | 14 - 15 | 1592 | 62 | ||
AK161024.1 | - | 14 - 15 | 1490 | 62 | ||
AK168019.1 | - | 2 - 3 | 138 | 62 | ||
BC031544.1 | - | 14 - 15 | 1571 | 3987201 | 62 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons |