Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hspa12a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK076668.1 | - | 2 - 3 | 297 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Mus musculus 1700 Probe spans exons |