Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Etv6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK078773.1 | - | 1 - 2 | 172 | 74 | |
AK154569.1 | - | 1 - 2 | 260 | 74 | ||
AK155150.1 | - | 1 - 2 | 445 | 74 | ||
AK157265.1 | - | 1 - 2 | 114 | 74 | ||
BC052163.1 | - | 1 - 2 | 133 | 6513848 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |