Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0300018769.mm.1 |
NM_001082484.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300018769.mm.1 |
NM_029721.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300018769.mm.1 |
TC0300002445.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300018764.mm.1 |
NM_001082484.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300018764.mm.1 |
NM_029721.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300018764.mm.1 |
TC0300002445.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300018765.mm.1 |
NM_001082484.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300018765.mm.1 |
NM_029721.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300018765.mm.1 |
TC0300002445.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300018767.mm.1 |
NM_001082484.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300018767.mm.1 |
NM_029721.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300018767.mm.1 |
TC0300002445.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300018768.mm.1 |
NM_001082484.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300018768.mm.1 |
NM_029721.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300018768.mm.1 |
TC0300002445.mm.1 |
MTA TC |