Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Neto1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_144946.4 | NP_659195.3 | 5 - 6 | 1445 | 65 | |
GenBank mRNA | AF448840.1 | - | 5 - 6 | 778 | 65 | |
AK032510.1 | - | 3 - 4 | 313 | 65 | ||
AK043710.1 | - | 5 - 6 | 781 | 65 | ||
AK081325.1 | - | 5 - 6 | 1446 | 65 | ||
AY138990.1 | - | 5 - 6 | 514 | 65 | ||
BC051145.1 | - | 5 - 6 | 805 | 6590345 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protease Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |