Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1100036178.mm.1 |
NM_001205236.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100036178.mm.1 |
NM_001205236.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100036178.mm.1 |
NM_133715.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100036178.mm.1 |
NM_133715.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100036178.mm.1 |
TC1100003871.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100036180.mm.1 |
NM_001205236.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100036180.mm.1 |
NM_001205236.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100036180.mm.1 |
NM_133715.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100036180.mm.1 |
NM_133715.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100036180.mm.1 |
TC1100003871.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100036179.mm.1 |
NM_001205236.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100036179.mm.1 |
NM_001205236.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100036179.mm.1 |
NM_133715.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100036179.mm.1 |
NM_133715.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100036179.mm.1 |
TC1100003871.mm.1 |
MTA TC |