Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cndp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001289531.1 | NP_001276460.1 | 4 - 5 | 619 | 78 | |
NM_023149.3 | NP_075638.2 | 5 - 6 | 587 | 78 | ||
GenBank mRNA | AB046738.1 | - | 5 - 6 | 528 | 78 | |
AK003779.1 | - | 5 - 6 | 508 | 78 | ||
AK131124.1 | - | 4 - 5 | 499 | 78 | ||
AK151013.1 | - | 5 - 6 | 509 | 78 | ||
AK152442.1 | - | 4 - 5 | 585 | 78 | ||
AK152729.1 | - | 4 - 5 | 557 | 78 | ||
AK153040.1 | - | 4 - 5 | 558 | 78 | ||
AK159129.1 | - | 4 - 5 | 585 | 78 | ||
AK159739.1 | - | 5 - 6 | 515 | 78 | ||
AK168029.1 | - | 5 - 6 | 526 | 78 | ||
AK168652.1 | - | 5 - 6 | 538 | 78 | ||
AK168856.1 | - | 3 - 4 | 276 | 78 | ||
AK172034.1 | - | 5 - 6 | 509 | 78 | ||
AK177956.1 | - | 1 - 2 | 44 | 78 | ||
AK189897.1 | - | 2 - 3 | 156 | 78 | ||
BC005532.1 | - | 5 - 6 | 518 | 3496319 | 78 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |