Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cndp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177450.4 | NP_803233.2 | 6 - 7 | 949 | 98 | |
GenBank mRNA | AK085308.1 | - | 6 - 7 | 939 | 98 | |
AK165706.1 | - | 6 - 7 | 901 | 98 | ||
BC043305.1 | - | 6 - 7 | 878 | 5040944 | 98 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |