Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mbp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025256.2 | NP_001020427.1 | 5 - 6 | 535 | 90 | |
NM_001025259.2 | NP_001020430.1 | 4 - 5 | 457 | 90 | ||
GenBank mRNA | AK005129.1 | - | 4 - 5 | 391 | 90 | |
AK079621.1 | - | 5 - 6 | 491 | 90 | ||
AK147966.1 | - | 4 - 5 | 405 | 90 | ||
AK187476.1 | - | 3 - 4 | 156 | 90 | ||
BB633793.1 | - | 5 - 6 | 491 | 90 | ||
M15060.1 | - | 5 - 6 | 455 | 90 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 3' Most Mus musculus Probe spans exons |