Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spire1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BC058669.1 | - | 7 - 8 | 775 | 93 | |
BC094375.1 | - | 7 - 8 | 828 | 30535423 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |